4-Aminopyrimidine 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H27367.MD
H27367.06
H27367.03
H27367.MDEA
2090
CZK
H27367.MD
H27367.06
H27367.03
4-Aminopyrimidine 98%
4-Aminopyrimidine
Purity: 98%
CAS No.: 591-54-8
Molecular Formula: C4H5N3
Formula Weight: 95.11
Melting Point: 147-152°
MDL No.: MFCD00006112
Vzorec:
C₄H₅N₃ Bod tání: 147…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006112 CAS číslo: 591-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...