2-Aminopyrimidine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyrimidin-2-ylamine, Pyrimidin-2-amine
M-Clarity™ quality level = MQ100
A78608-100G
A78608-500G
SIALA78608-100GEA
1340
CZK
SIALA78608-100G
SIALA78608-500G
2-Aminopyrimidine, Sigma-Aldrich®
2-Aminopyrimidine
Vzorec:
C₄H₅N₃ MW: 95,1 g/mol Bod varu: 275 °C (1013 hPa) Bod tání: 123…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006089 CAS číslo: 109-12-6 EINECS: 203-648-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...