Tisk…

2-Aminopyrimidine 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Pyrimidin-2-ylamine, Pyrimidin-2-amine

B24594.22 B24594.36
B24594.22EA 1380 CZK
B24594.22 B24594.36
2-Aminopyrimidine 98%
2-Aminopyrimidine

Purity: 98% CAS No.: 109-12-6 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 123-127° Boiling Point: 159°/186mm MDL No.: MFCD00006089

Vzorec: C₄H₅N₃
MW: 95,1 g/mol
Bod varu: 159 °C (186 mmHg)
Bod tání: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00006089
CAS číslo: 109-12-6
EINECS: 203-648-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR