Tisk…

2,4,5-Trichloropyrimidine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L19506.03 L19506.06
L19506.03EA 1950 CZK
L19506.03 L19506.06
2,4,5-Trichloropyrimidine ≥98%
2,4,5-Trichloropyrimidine

Purity: 98% CAS No.: 5750-76-5 Molecular Formula: C4HCl3N2 Formula Weight: 183.43 Flash Point: 110°(230°F) Refractive Index: 1.5730 Boiling Point: 84°/1mm Beilstein Reference: 4449 UN No.: 1760 MDL No.: MFCD03788200

Vzorec: C₄HCl₃N₂
Bod varu: 84 °C (1 mmHg)
Hustota: 1,600
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03788200
CAS číslo: 5750-76-5
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR