Tisk…

4-Amino-2-methylpyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Amino-2-picoline, 2-Methyl-4-pyridylamine, 2-Methylpyridin-4-amine

M-Clarity™ quality level = MQ100

662712-1G 662712-250MG
SIAL662712-1GEA 10990 CZK
SIAL662712-1G SIAL662712-250MG
4-Amino-2-methylpyridine, Sigma-Aldrich®
4-Amino-2-methylpyridine
Vzorec: C₆H₈N₂
MW: 108,14 g/mol
MDL Number: MFCD00186509
CAS číslo: 18437-58-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR