5-Amino-2-methylpyridine 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
5-Amino-2-picoline, 6-Methylpyridin-3-amine, 3Amino6methylpyridine, 6-Methyl-3-pyridinamine, 3Amino6picoline
H27431.03
H27431.MD
H27431.03EA
6180
CZK
H27431.03
H27431.MD
5-Amino-2-methylpyridine 97%
5-Amino-2-methylpyridine
Purity: 97%
CAS No.: 3430-14-6
Molecular Formula: C6H8N2
Formula Weight: 108.14
Odour: Characteristic
Melting Point: 95-99°
UN No.: 2811
MDL No.: MFCD00833389
Vzorec:
C₆H₈N₂ MW: 108,14 g/mol Bod tání: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00833389 CAS číslo: 3430-14-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...