Tisk…

3-Amino-4-methylpyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

697141-25G 697141-5G
SIAL697141-25GEA 8330 CZK
SIAL697141-25G SIAL697141-5G
3-Amino-4-methylpyridine, Sigma-Aldrich®
3-Amino-4-methylpyridine
Vzorec: C₆H₈N₂
MW: 108,14 g/mol
MDL Number: MFCD00128871
CAS číslo: 3430-27-1
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR