Tisk…

2-Amino-6-methylpyridine 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline

A10444.22 A10444.36
A10444.22EA 1400 CZK
A10444.22 A10444.36
2-Amino-6-methylpyridine 99%
2-Amino-6-methylpyridine

Purity: 99% CAS No.: 1824-81-3 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 103°(217°F) Odour: Unpleasant Boiling Point: 208-209° Beilstein Reference: 107048 UN No.: 2811 MDL No.: MFCD00006331

Vzorec: C₆H₈N₂
MW: 108,14 g/mol
Bod varu: 208…209 °C
Bod tání: 41…45 °C
Bod vzplanutí: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006331
CAS číslo: 1824-81-3
EINECS: 217-360-1
UN: 2811
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR