2-Amino-3-methylpyridine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline
M-Clarity™ quality level = MQ100
A75633-100G
A75633-500G
SIALA75633-100GEA
2370
CZK
SIALA75633-100G
SIALA75633-500G
2-Amino-3-methylpyridine, Sigma-Aldrich®
2-Amino-3-methylpyridine
Vzorec:
C₆H₈N₂ MW: 108.14 Bod varu: 221.1 Bod tání: 29…32 °C Hustota: 1.073 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006320 CAS číslo: 1603-40-3 EINECS: 216-501-4 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...