4-Acetylpyridine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Methyl 4-pyridyl ketone
A12015.14
A12015.22
A12015.14EA
1320
CZK
A12015.14
A12015.22
4-Acetylpyridine ≥98%
4-Acetylpyridine
Purity: 98%
CAS No.: 1122-54-9
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 104°(219°F)
Refractive Index: 1.5350
Melting Point: 14-16°
Boiling Point: 210-212°
Beilstein Reference: 107629
MDL No.: MFCD00006433
Vzorec:
C₇H₇NO Bod varu: 210…212 °C Bod tání: 14…16 °C Hustota: 1,103 Bod vzplanutí: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006433 CAS číslo: 1122-54-9 EINECS: 214-350-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...