Tisk…

3-Acetylpyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone

M-Clarity™ quality level = MQ300

W342408-100G-K W342408-10KG-K W342408-1KG-K W342408-5KG-K W342408-SAMPLE
SIALW342408-100G-KEA 4130 CZK
SIALW342408-100G-K SIALW342408-10KG-K SIALW342408-1KG-K SIALW342408-5KG-K SIALW342408-SAMPLE
3-Acetylpyridine, Sigma-Aldrich®
3-Acetylpyridine
Vzorec: C₇H₇NO
Bod varu: 220 °C (1013 hPa)
Hustota: 1,1065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS číslo: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III
Merck Index: 12,06195

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR