2-Acetylpyridine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-(Pyridin-2-yl)ethanone, Methyl 2-pyridyl ketone
M-Clarity™ quality level = MQ300
W325104-100G-K
W325104-1KG-K
W325104-25KG-K
W325104-5KG-K
SIALW325104-100G-KEA
2100
CZK
SIALW325104-100G-K
SIALW325104-1KG-K
SIALW325104-25KG-K
SIALW325104-5KG-K
2-Acetylpyridine, Sigma-Aldrich®
2-Acetylpyridine
Vzorec:
C₇H₇NO MW: 121,14 g/mol Bod varu: 192,3 °C (1013 hPa) Bod tání: ∼10 °C Hustota: 1,0818 g/cm³ (20 °C) Bod vzplanutí: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006303 CAS číslo: 1122-62-9 EINECS: 214-355-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...