Tisk…

2-Amino-5-bromo-4-methylpyridine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline

44623.06 44623.14
44623.06EA 3300 CZK
44623.06 44623.14
2-Amino-5-bromo-4-methylpyridine ≥97%
2-Amino-5-bromo-4-methylpyridine

Purity: 97% CAS No.: 98198-48-2 Molecular Formula: C6H7BrN2 Formula Weight: 187.04 Physical Form: Powder Appearance: Cream Melting Point: 148-151° MDL No.: MFCD03427660

Vzorec: C₆H₇BrN₂
MW: 187.04
Bod tání: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD03427660
CAS číslo: 98198-48-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR