4-Bromo-2-methylpyridine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H27884.MD
H27884.03
H27884.MDEA
1180
CZK
H27884.MD
H27884.03
4-Bromo-2-methylpyridine ≥97%
4-Bromo-2-methylpyridine
Purity: 97%
CAS No.: 22282-99-1
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point: 79°(174°F)
Refractive Index: 1.556
Odour: Characteristic
Boiling Point: 76°/14mm
MDL No.: MFCD03788196
Vzorec:
C₆H₆BrN Bod varu: 76 °C (14 mmHg) Hustota: 1,45 Bod vzplanutí: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03788196 CAS číslo: 22282-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...