Tisk…

2-Bromo-6-methylpyridine, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 6-Bromo-2-picoline, 2-Bromo-6-picoline

M-Clarity™ quality level = MQ100

429112-10G 429112-1G
SIAL429112-10GEA 4410 CZK
SIAL429112-10G SIAL429112-1G
2-Bromo-6-methylpyridine, Sigma-Aldrich®
2-Bromo-6-methylpyridine
Vzorec: C₆H₆BrN
MW: 172,02 g/mol
MDL Number: MFCD00040743
CAS číslo: 5315-25-3
EINECS: 226-173-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR