2-Pyridinethiol 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Pyridine-2-thiol, 2-Mercaptopyridine
A11741.06
A11741.14
A11741.22
A11741.06EA
1120
CZK
A11741.06
A11741.14
A11741.22
2-Pyridinethiol 98%
2-Pyridinethiol
Purity: 98%
CAS No.: 2637-34-5
Molecular Formula: C5H5NS
Formula Weight: 111.17
Storage Temperature: 2°C to 8°C
Melting Point: 124-130°
Beilstein Reference: 105758
MDL No.: MFCD00006285
Vzorec:
C₅H₅NS MW: 111,17 g/mol Bod varu: 259 °C (1013 hPa) Bod tání: 124…130 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006285 CAS číslo: 2637-34-5 EINECS: 220-131-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...