3-Hydroxy-2-pyridone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,3-Pyridinediol, 3-Hydroxy-2(1H)-pyridinone, 3-Hydroxy-2-pyridone, 2,3-Dihydroxypyridine, Pyridine-2,3-diol
M-Clarity™ quality level = MQ100
122505-100G
122505-25G
SIAL122505-100GEA
10340
CZK
SIAL122505-100G
SIAL122505-25G
3-Hydroxy-2-pyridone, Sigma-Aldrich®
3-Hydroxy-2-pyridone
Vzorec:
C₅H₅NO₂ MW: 111,1 g/mol Bod varu: 315 °C (1013 hPa) Bod tání: 244…246 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006271 CAS číslo: 16867-04-2 EINECS: 240-887-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...