Tisk…

4-Pyridone ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A14586.14 A14586.22 A14586.36
A14586.14EA 1630 CZK
A14586.14 A14586.22 A14586.36
4-Pyridone ≥97%
4-Pyridone

Purity: 97% CAS No.: 626-64-2 Molecular Formula: C5H5NO Formula Weight: 95.10 Melting Point: 142-148° Boiling Point: 230-235°/12mm MDL No.: MFCD00040458

Vzorec: C₅H₅NO
Bod varu: 230…235 °C (12 mmHg)
Bod tání: 142…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00006419
CAS číslo: 626-64-2
EINECS: 210-958-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR