3-Pyridone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol
A13910.18
A13910.22
A13910.36
A13910.18EA
1250
CZK
A13910.18
A13910.22
A13910.36
3-Pyridone ≥98%
3-Pyridone
Purity: 98%
CAS No.: 109-00-2
Molecular Formula: C5H5NO
Formula Weight: 95.10
Flash Point: 173°(343°F)
Melting Point: 125-128°
Boiling Point: 180°/22mm
Beilstein Reference: 105699
MDL No.: MFCD00006378
Vzorec:
C₅H₅NO MW: 95,1 g/mol Bod varu: 180 °C (22 mmHg) Bod tání: 125…128 °C Hustota: 1,2 g/cm³ (–70 °C) Bod vzplanutí: 173 °C (343 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006378 CAS číslo: 109-00-2 EINECS: 203-637-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...