2-Methoxypyridine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
M25406-100G
M25406-25G
SIALM25406-100GEA
2570
CZK
SIALM25406-100G
SIALM25406-25G
2-Methoxypyridine, Sigma-Aldrich®
2-Methoxypyridine
Vzorec:
C₆H₇NO Bod varu: 142 °C (1013 hPa) Hustota: 1,064 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006262 CAS číslo: 1628-89-3 EINECS: 216-623-8 UN: 3271 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...