Pyridine-N-oxide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyridine 1-oxide, Pyridine N-oxide
M-Clarity™ quality level = MQ100
131652-100G
131652-25G
SIAL131652-100GEA
2530
CZK
SIAL131652-100G
SIAL131652-25G
Pyridine-N-oxide, Sigma-Aldrich®
Pyridine-N-oxide
Vzorec:
C₅H₅NO MW: 95,1 g/mol Bod varu: 270 °C (1013 hPa) Bod tání: 61…64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006194 CAS číslo: 694-59-7 EINECS: 211-774-6 Merck Index: 12,08155 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...