Tisk…

Pyridine-N-oxide 95%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Pyridine 1-oxide, Pyridine N-oxide

A10419.14 A10419.22 A10419.36
A10419.14EA 841 CZK
A10419.14 A10419.22 A10419.36
Pyridine-N-oxide 95%
Pyridine-N-oxide

Purity: 95% CAS No.: 694-59-7 Molecular Formula: C5H5NO Formula Weight: 95.10 Flash Point: 143°(289°F) Melting Point: 61-66° Boiling Point: 270° Beilstein Reference: 105257 Merck Reference: 14,7972 MDL No.: MFCD00006194

Vzorec: C₅H₅NO
MW: 95,1 g/mol
Bod varu: 270 °C
Bod tání: 61…66 °C
Bod vzplanutí: 143 °C (289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006194
CAS číslo: 694-59-7
EINECS: 211-774-6
Merck Index: 14,7972

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR