2-(Dimethylamino)pyridine 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L02513.06
L02513.14
L02513.06EA
1090
CZK
L02513.06
L02513.14
2-(Dimethylamino)pyridine 97%
2-(Dimethylamino)pyridine
Purity: 97%
CAS No.: 5683-33-0
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 80°(176°F)
Refractive Index: 1.5590
Boiling Point: 190-192°
Beilstein Reference: 110353
MDL No.: MFCD00006261
Vzorec:
C₇H₁₀N₂ Bod varu: 190…192 °C Hustota: 1,008 Bod vzplanutí: 80 °C (176 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006261 CAS číslo: 5683-33-0 EINECS: 227-147-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...