2-Iodo-3-methoxypyridine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L19868.03
L19868.06
L19868.03EA
810
CZK
L19868.03
L19868.06
2-Iodo-3-methoxypyridine ≥97%
2-Iodo-3-methoxypyridine
Purity: 97%
CAS No.: 93560-55-5
Molecular Formula: C6H6INO
Formula Weight: 235.03
Melting Point: 57-61°
Beilstein Reference: 115569
Vzorec:
C₆H₆INO Bod tání: 57…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234181 CAS číslo: 93560-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...