Tisk…

2-Amino-3-iodo-5-nitropyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Iodo-5-nitropyridin-2-amine, 3-Iodo-5-nitro-2-pyridylamine

M-Clarity™ quality level = MQ100

694738-1G 694738-5G
SIAL694738-1GEA 0 CZK
SIAL694738-1G SIAL694738-5G
2-Amino-3-iodo-5-nitropyridine, Sigma-Aldrich®
2-Amino-3-iodo-5-nitropyridine
Vzorec: C₅H₄IN₃O₂
MW: 265,01 g/mol
MDL Number: MFCD07368863
CAS číslo: 25391-56-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR