Tisk…

2,5-Diiodopyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

652733-25G 652733-5G
SIAL652733-25GEA 7690 CZK
SIAL652733-25G SIAL652733-5G
2,5-Diiodopyridine, Sigma-Aldrich®
2,5-Diiodopyridine
Vzorec: C₅H₃I₂N
MDL Number: MFCD00234028
CAS číslo: 116195-81-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR