Tisk…

2-Amino-5-fluoropyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

518689-1G
SIAL518689-1GEA 3740 CZK
SIAL518689-1G
2-Amino-5-fluoropyridine, Sigma-Aldrich®
2-Amino-5-fluoropyridine
Vzorec: C₅H₅FN₂
MW: 112.11
MDL Number: MFCD01861120
CAS číslo: 21717-96-4
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR