Tisk…

3-Fluoro-4-iodopyridine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

685216-1G 685216-5G
SIAL685216-1GEA 6260 CZK
SIAL685216-1G SIAL685216-5G
3-Fluoro-4-iodopyridine, Sigma-Aldrich®
3-Fluoro-4-iodopyridine
Vzorec: C₅H₃FIN
MW: 222,99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03092927
CAS číslo: 22282-75-3
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR