Tisk…
Safety shoes, lace-up, Run-R 210, Heckel
Dodavatel: Uvex
Run-R
UVEX6264347EA 2910 CZK
UVEX6264347 UVEX6264348 UVEX6264337 UVEX6264338 UVEX6264340 UVEX6264341 UVEX6264342 UVEX6264344 UVEX6264346
Safety shoes, lace-up, Run-R 210, Heckel
Obuv
Metal-free, flexible and lightweight safety shoe made from microfibre/TPU film upper and a double density PU2D sole, highly slip resistant and resistant to oils and hydrocarbons.
  • 3D polyester mesh lining
  • Anatomically shaped hygienic insole
  • Shock absorber heel and front
  • Metal-free protective toecap (200 J)
  • Metal-free puncture resistant midsole
Suitable for light industry, warehousing and logistics, manufacturing, dry environments.
Colour: Black/orange
Weight: 490 g
Certifikace: EN ISO 20345:2011; S1P, SRC