2-Fluoropyridine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
F15250-100G
F15250-10G
SIALF15250-100GEA
7960
CZK
SIALF15250-100G
SIALF15250-10G
2-Fluoropyridine, Sigma-Aldrich®
2-Fluoropyridine
Vzorec:
C₅H₄FN MW: 97,09 g/mol Bod varu: 126 °C (1013 hPa) Hustota: 1,129 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006224 CAS číslo: 372-48-5 EINECS: 206-757-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...