Isonicotinamide ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A18190.18
A18190.22
A18190.30
A18190.36
A18190.18EA
1330
CZK
A18190.18
A18190.22
A18190.30
A18190.36
Isonicotinamide ≥99%
Isonicotinamide
Purity: 99%
CAS No.: 1453-82-3
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Melting Point: 155-158°
Beilstein Reference: 2173
Vzorec:
C₆H₆N₂O Bod varu: 334 °C (1013 hPa) Bod tání: 155…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006432 CAS číslo: 1453-82-3 EINECS: 215-926-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...