Tisk…

Pyridine-2,6-dicarboxylic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA

A12263.14 A12263.22 A12263.36
A12263.14EA 855 CZK
A12263.14 A12263.22 A12263.36
Pyridine-2,6-dicarboxylic acid ≥98%
Pyridine-2,6-dicarboxylic acid

Purity: 98% CAS No.: 499-83-2 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Flash Point: 188°(370°F) Melting Point: ca 245° dec. Beilstein Reference: 131629 MDL No.: MFCD00006299

Vzorec: C₇H₅NO₄
MW: 167.12
Bod varu: 357
Bod tání: ∼ 245 °C (rozkl.)
Bod vzplanutí: 188 °C (370 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS číslo: 499-83-2
EINECS: 207-894-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR