Tisk…

6-Hydroxynicotinic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid

A15802.06 A15802.14 A15802.22
A15802.06EA 766 CZK
A15802.06 A15802.14 A15802.22
6-Hydroxynicotinic acid ≥98%
6-Hydroxynicotinic acid

Purity: 98% CAS No.: 5006-66-6 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Melting Point: 314-316° Beilstein Reference: 472182

Vzorec: C₆H₅NO₃
MW: 139,11 g/mol
Bod tání: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006277
CAS číslo: 5006-66-6
EINECS: 225-682-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR