Tisk…

2-Hydroxynicotinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Hydroxypyridine-3-carboxylic acid, 1,2-Dihydro-2-oxonicotinic acid, 2-Hydroxy-3-pyridinecarboxylic acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic acid

M-Clarity™ quality level = MQ100

251054-100G 251054-25G
SIAL251054-100GEA 5100 CZK
SIAL251054-100G SIAL251054-25G
2-Hydroxynicotinic acid, Sigma-Aldrich®
2-Hydroxynicotinic acid
Vzorec: C₆H₅NO₃
MW: 139,11 g/mol
MDL Number: MFCD00010100
CAS číslo: 609-71-2
EINECS: 210-198-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR