Tisk…

2-Aminonicotinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid

M-Clarity™ quality level = MQ100

A68300-25G A68300-5G
SIALA68300-25GEA 5070 CZK
SIALA68300-25G SIALA68300-5G
2-Aminonicotinic acid, Sigma-Aldrich®
2-Aminonicotinic acid
Vzorec: C₆H₆N₂O₂
MW: 138,13 g/mol
Bod tání: 297 °C
Hustota: 1,545 g/cm³ (20 °C)
MDL Number: MFCD00006318
CAS číslo: 5345-47-1
EINECS: 226-296-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR