Isonicotinic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
A18109.0E
A18109.22
A18109.36
A18109.0EEA
15790
CZK
A18109.0E
A18109.22
A18109.36
Isonicotinic acid ≥99%
Isonicotinic acid
Purity: 99%
CAS No.: 55-22-1
Molecular Formula: C6H5NO2
Formula Weight: 123.11
pH: 3.0-4.0
Melting Point: ca 310° subl.
Boiling Point: 260°/15mm
Beilstein Reference: 109599
Merck Reference: 14,5187
MDL No.: MFCD00006429
Vzorec:
C₆H₅NO₂ MW: 123,11 g/mol Bod varu: 260 °C (15 mmHg) Bod tání: ∼ 310 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00006429 CAS číslo: 55-22-1 EINECS: 200-228-2 Merck Index: 14,5187 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...