Tisk…

Picolinic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid

A14749.14 A14749.22 A14749.36
A14749.14EA 779 CZK
A14749.14 A14749.22 A14749.36
Picolinic acid ≥99%
Picolinic acid

Purity: 99% CAS No.: 98-98-6 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 139°(232°F) Odour: Odourless Beilstein Reference: 109595 Merck Reference: 14,7403 MDL No.: MFCD00006293

Vzorec: C₆H₅NO₂
MW: 123.11
Bod varu: 293
Bod tání: 135…139 °C
Hustota: 1.49 g/cm³ (20 °C)
Bod vzplanutí: 139 °C (232 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS číslo: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR