Tisk…

Methylnikotinát, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate

M-Clarity™ quality level = MQ100

M59203-5G M59203-100G M59203-500G
SIALM59203-5GEA 465 CZK
SIALM59203-5G SIALM59203-100G SIALM59203-500G
Methylnikotinát, Sigma-Aldrich®
Methylnikotinát
Vzorec: C₇H₇NO₂
MW: 137,14 g/mol
Bod varu: 209 °C (1013 hPa)
Bod tání: 39 °C
Hustota: 1,0048 g/cm³ (20 °C)
Bod vzplanutí: 95 °C
MDL Number: MFCD00006388
CAS číslo: 93-60-7
EINECS: 202-261-8
Merck Index: 12,06174

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR