Methylnikotinát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate
M-Clarity™ quality level = MQ100
72420-100G
72420-500G
SIAL72420-100GEA
1220
CZK
SIAL72420-100G
SIAL72420-500G
Methylnikotinát, Sigma-Aldrich®
Methylnikotinát
Vzorec:
C₇H₇NO₂ MW: 137,14 g/mol Bod varu: 209 °C (1013 hPa) Bod tání: 39 °C Hustota: 1,0048 g/cm³ (20 °C) Bod vzplanutí: 95 °C |
MDL Number:
MFCD00006388 CAS číslo: 93-60-7 EINECS: 202-261-8 Merck Index: 12,06174 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...