Ethyl nicotinate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Pyridinecarboxylic acid ethyl ester, Ethyl 3-pyridinecarboxylate, Pyridine-3-carboxylic acid ethyl ester, 3-Picolinic acid ethyl ester, Nicotinic acid ethyl ester
L03215.22
L03215.36
L03215.22EA
1030
CZK
L03215.22
L03215.36
Ethyl nicotinate ≥99%
Ethyl nicotinate
Purity: 99%
CAS No.: 614-18-6
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Flash Point: 93°(199°F)
Refractive Index: 1.5030
Odour: Characteristic
Melting Point: 8-9°
Boiling Point: 223-224°
Beilstein Reference: 122937
MDL No.: MFCD00006389
Vzorec:
C₈H₉NO₂ Bod varu: 223…224 °C Bod tání: 8…9 °C Hustota: 1,107 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006389 CAS číslo: 614-18-6 EINECS: 210-370-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...