Nicotinonitrile, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
3-Pyridinecarbonitrile, 3-Cyanopyridine, Pyridine-3-carbonitrile
M-Clarity™ quality level = MQ100
C94807-100G
C94807-5G
SIALC94807-100GEA
1010
CZK
SIALC94807-100G
SIALC94807-5G
Nicotinonitrile, Sigma-Aldrich®
Nicotinonitrile
Vzorec:
C₆H₄N₂ MW: 104.11 Bod varu: 206.2 Bod tání: 48…51 °C Hustota: 1.159 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00087130 CAS číslo: 100-54-9 EINECS: 202-863-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...