Nicotinonitrile ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Pyridinecarbonitrile, 3-Cyanopyridine, Pyridine-3-carbonitrile
A14850.0E
A14850.22
A14850.36
A14850.0EEA
14760
CZK
A14850.0E
A14850.22
A14850.36
Nicotinonitrile ≥98%
Nicotinonitrile
Purity: 98%
CAS No.: 100-54-9
Molecular Formula: C6H4N2
Formula Weight: 104.11
Flash Point: 84°(183°F)
Odour: Pungent
Boiling Point: 206-208°
Beilstein Reference: 107711
MDL No.: MFCD00006372
Vzorec:
C₆H₄N₂ MW: 104.11 Bod varu: 206…208 °C Bod tání: 48…51 °C Hustota: 1.16 Bod vzplanutí: 84 °C (183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00087130 CAS číslo: 100-54-9 EINECS: 202-863-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...