3-Chloro-2-hydroxypyridine ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Chloro-2-pyridinol
L20027.03
L20027.06
L20027.14
L20027.03EA
889
CZK
L20027.03
L20027.06
L20027.14
3-Chloro-2-hydroxypyridine ≥97%
3-Chloro-2-hydroxypyridine
Purity: 97%
CAS No.: 13466-35-8
Molecular Formula: C5H4ClNO
Formula Weight: 129.55
Melting Point: 180-182°
Beilstein Reference: 109845
Vzorec:
C₅H₄ClNO MW: 129,55 g/mol Bod tání: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646109 CAS číslo: 13466-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...