2-Chloro-5-fluoropyridine 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H27462.06
H27462.03
H27462.06EA
2700
CZK
H27462.06
H27462.03
2-Chloro-5-fluoropyridine 95%
2-Chloro-5-fluoropyridine
Purity: 95%
CAS No.: 31301-51-6
Molecular Formula: C5H3ClFN
Formula Weight: 131.54
Flash Point: 42°(108°F)
Refractive Index: 1.503
Melting Point: 19°
Boiling Point: 74°/38mm
UN No.: 2929
MDL No.: MFCD03095332
Vzorec:
C₅H₃ClFN Bod varu: 74 °C (38 mmHg) Bod tání: 19 °C Hustota: 1,297 Bod vzplanutí: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03095332 CAS číslo: 31301-51-6 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...