2-Chloronicotinic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid
M-Clarity™ quality level = MQ100
150339-10G
150339-50G
SIAL150339-10GEA
0
CZK
SIAL150339-10G
SIAL150339-50G
2-Chloronicotinic acid, Sigma-Aldrich®
2-Chloronicotinic acid
Vzorec:
C₆H₄ClNO₂ MW: 157,56 g/mol Bod varu: 284 °C (1013 hPa) Bod tání: 181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006236 CAS číslo: 2942-59-8 EINECS: 220-937-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...