Tisk…

6-Chloronicotinamide ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Chloropyridine-3-carboxamide

B21922.03 B21922.06 B21922.14
B21922.03EA 639 CZK
B21922.03 B21922.06 B21922.14
6-Chloronicotinamide ≥98%
6-Chloronicotinamide

Purity: 98% CAS No.: 6271-78-9 Molecular Formula: C6H5ClN2O Formula Weight: 156.57 Melting Point: 214-216° Beilstein Reference: 116043

Vzorec: C₆H₅ClN₂O
MW: 156,57 g/mol
Bod tání: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00006242
CAS číslo: 6271-78-9
EINECS: 228-456-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR