Tisk…

2-Bromo-3-methoxypyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

649716-10G 649716-1G
SIAL649716-10GEA 10880 CZK
SIAL649716-10G SIAL649716-1G
2-Bromo-3-methoxypyridine, Sigma-Aldrich®
2-Bromo-3-methoxypyridine
Vzorec: C₆H₆BrNO
MW: 188,02 g/mol
MDL Number: MFCD01570896
CAS číslo: 24100-18-3
EINECS: 246-017-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR