Tisk…

5-Amino-2-bromopyridine, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 6-Bromopyridin-3-amine, 2-Bromopyridin-5-amine, 3-Amino-6-bromopyridine, 2-Bromo-5-pyridylamine

M-Clarity™ quality level = MQ200

552844-1G 552844-5G
SIAL552844-1GEA 1580 CZK
SIAL552844-1G SIAL552844-5G
5-Amino-2-bromopyridine, Sigma-Aldrich®
5-Amino-2-bromopyridine
Vzorec: C₅H₅BrN₂
MW: 173,01 g/mol
MDL Number: MFCD01646116
CAS číslo: 13534-97-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR