3-Bromo-5-fluoropyridine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
646296-1G
646296-5G
SIAL646296-1GEA
0
CZK
SIAL646296-1G
SIAL646296-5G
3-Bromo-5-fluoropyridine, Sigma-Aldrich®
3-Bromo-5-fluoropyridine
Vzorec:
C₅H₃BrFN MW: 175,99 g/mol |
MDL Number:
MFCD04112555 CAS číslo: 407-20-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...