3-Bromo-5-fluoropyridine 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H25766.03
H25766.06
H25766.03EA
1180
CZK
H25766.03
H25766.06
3-Bromo-5-fluoropyridine 97%
3-Bromo-5-fluoropyridine
Purity: 99%
CAS No.: 407-20-5
Molecular Formula: C5H3BrFN
Formula Weight: 175.99
Flash Point: 64°(148°F)
Odour: Characteristic
UN No.: 2811
MDL No.: MFCD04112555
Vzorec:
C₅H₃BrFN Bod tání: 24…28 °C Bod vzplanutí: 64 °C (148 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04112555 CAS číslo: 407-20-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...